missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methyl-2-(methylthio)propionate, 98%, Thermo Scientific™

Catalog Number p-7026547
Click to view available options
Quantity:
2.5 g
10 g
This item is not returnable. View return policy
49773-24-2
C7H14O2S
162.247
MFCD00053099
JLURSSXMQSLZQL-UHFFFAOYSA-N
ethyl 2-methyl-2-methylthio propionate, propanoic acid, 2-methyl-2-methylthio-, ethyl ester, ethyl 2-methyl-2-methylthio propanoate, ethyl 2-methyl-2-methylsulfanyl propanoate, acmc-20al0x, ethyl 2-methylthio isobutyrate, 2-methyl-2-methylthio propionic acid ethyl ester, 2-methyl-2-methylthio-propanoic acid ethyl ester
3016499
ethyl 2-methyl-2-methylsulfanylpropanoate
CCOC(=O)C(C)(C)SC
This item is not returnable. View return policy
49773-24-2
C7H14O2S
162.247
MFCD00053099
JLURSSXMQSLZQL-UHFFFAOYSA-N
ethyl 2-methyl-2-methylthio propionate, propanoic acid, 2-methyl-2-methylthio-, ethyl ester, ethyl 2-methyl-2-methylthio propanoate, ethyl 2-methyl-2-methylsulfanyl propanoate, acmc-20al0x, ethyl 2-methylthio isobutyrate, 2-methyl-2-methylthio propionic acid ethyl ester, 2-methyl-2-methylthio-propanoic acid ethyl ester
3016499
ethyl 2-methyl-2-methylsulfanylpropanoate
CCOC(=O)C(C)(C)SC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 2.5 g
Formula Weight 162.25
Percent Purity 98%
Chemical Name or Material Ethyl 2-methyl-2-(methylthio)propionate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'