Ethyl 2-mercaptoacetate, 99%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 118670025

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 623-51-8 Weight: 4.00000kg Transport: UN number : 1228 Chem class : 2 Pack group : III


Packaging Glass bottle
Quantity 2.5kg

Product Code. 10483055

Quantity Price
1 £ 336.7 / 2.50kg
EU Stock 1
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethyl 2-mercaptoacetate
Name Note 99%
CAS 623-51-8
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C4H8O2S
Linear Formula HSCH2CO2C2H5
Formula Weight 120.17
MDL Number MFCD00004874
Synonym Ethyl thioglycolate

Specification

Formula Weight 120.17
Assay ≥98.5%
Physical Form Liquid
Quantity 2.5kg
Color Clear colorless
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 210-800-3
Linear Formula HSCH2CO2C2H5
CAS 623-51-8
Infrared Spectrum Authentic
MDL Number MFCD00004874
Refractive Index 1.4560 to 1.4590 (20°C, 589nm)
Assay Percent Range 99%
RTECS Number AI6650000
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Packaging Glass bottle
Name Note 99%
Health Hazard 1 GHS Signal Word: Danger
Density 1.0900g/mL
Molecular Formula C4H8O2S
Synonym Ethyl thioglycolate
TSCA TSCA
Chemical Name or Material Ethyl 2-mercaptoacetate
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.