missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl (2-fluorobenzoyl)acetate, 97%

Brand:  Alfa Aesar™ H61204.06

Code : NEW

Additional Details : CAS Number : 1479-24-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15467708

  • £57.45 / 5g


Estimated Shipment date
from Supplier 24-04-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl (2-fluorobenzoyl)acetate
210.2
2-Fluorobenzoylacetic acid ethyl ester, 97%
1479-24-9
MFCD00000321

Specifications

Ethyl (2-fluorobenzoyl)acetate
1.164g/mL
97%
210.2
2-Fluorobenzoylacetic acid ethyl ester, 97%
1479-24-9
Liquid
1.5160
MFCD00000321
5g
Videos
SDS