missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-ethylacetoacetate, 95%

Brand:  Alfa Aesar™ B22295.22

Code : NEW

Additional Details : CAS Number : 607-97-6 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15403247

  • £207.00 / 100g

Estimated Shipment: 12-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

607-97-6
158.197
OKANYBNORCUPKZ-UHFFFAOYSA-N
79065
CCC(C(=O)C)C(=O)OCC
C8H14O3
MFCD00039898
ethyl 2-ethylacetoacetate, ethyl 2-acetylbutyrate, butanoic acid, 2-ethyl-3-oxo-, ethyl ester, 2-ethylacetoacetic acid ethyl ester, ethyl 2-ethyl-3-ketobutyrate, ethyl alpha-ethylacetoacetate, ethyl-2-ethylacetoacetate, 2-ethyl-3-oxo-butyric acid ethyl ester, ethyl .alpha.-acetylbutyrate, ethyl 2-ethyl-3-oxo-butanoate
ethyl 2-ethyl-3-oxobutanoate

Specifications

Ethyl 2-ethylacetoacetate
77°C (170°F)
83°C to 85°C (10mmHg)
607-97-6
MFCD00039898
ethyl 2-ethylacetoacetate, ethyl 2-acetylbutyrate, butanoic acid, 2-ethyl-3-oxo-, ethyl ester, 2-ethylacetoacetic acid ethyl ester, ethyl 2-ethyl-3-ketobutyrate, ethyl alpha-ethylacetoacetate, ethyl-2-ethylacetoacetate, 2-ethyl-3-oxo-butyric acid ethyl ester, ethyl .alpha.-acetylbutyrate, ethyl 2-ethyl-3-oxo-butanoate
CCC(C(=O)C)C(=O)OCC
158.197
158.2
1.009
1.422
100g
C8H14O3
636286
OKANYBNORCUPKZ-UHFFFAOYSA-N
ethyl 2-ethyl-3-oxobutanoate
79065
95%
Videos
SDS Documents
Certificates