missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-cyclohexylthiazolidine-4-carboxylate, 97%

Brand:  Alfa Aesar™ H62854.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459308

  • £119.78 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-cyclohexylthiazolidine-4-carboxylate
243.37
MFCD26967919
2-Cyclohexylthiazolidine-4-carboxylic acid ethyl ester

Specifications

Ethyl 2-cyclohexylthiazolidine-4-carboxylate
Solid
243.37
2-Cyclohexylthiazolidine-4-carboxylic acid ethyl ester
1g
97%
MFCD26967919
Videos
SDS