missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-cyanoacetoacetate, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
Description
It is used as a pharmaceutical intermediate. It is also used in the synthesis of fluorine-containing alkyl 2-cyano-3-oxocarboxylates and also in the syntheses of pyrazolo [1, 2-a] pyrazole and pyrazolo [5, 1-b] [1, 3] oxazine derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 2 g |
UN Number | UN3276 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 155.16 |
Physical Form | Liquid |
Chemical Name or Material | Ethyl 2-cyanoacetoacetate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'