missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-bromopropionate, 98+%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
250 g
500 g
Description
It’s an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It’s an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.
Solubility
Immiscible in water. Soluble in alcohol and ether.
Notes
Store in dark and away from oxidizing agents and acids.
It’s an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.
Solubility
Immiscible in water. Soluble in alcohol and ether.
Notes
Store in dark and away from oxidizing agents and acids.
Spécification
Density | 1.391 |
Boiling Point | 158°C to 160°C |
Flash Point | 51°C (123°F) |
Refractive Index | 1.446 |
Quantity | 50 g |
UN Number | UN1992 |
Beilstein | 773920 |
Merck Index | 14,3772 |
Solubility Information | Immiscible in water. Soluble in alcohol and ether. |
Formula Weight | 181.03 |
Afficher plus |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'