missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-bromo-4-(trifluoromethyl)thiazole-5-carboxylate, 97%

Brand:  Alfa Aesar™ H63546.06

Code : NEW

Additional Details : CAS Number : 72850-79-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15488356

  • £239.56 / 5g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-bromo-4-(trifluoromethyl)thiazole-5-carboxylate
304.08
2-Bromo-4-(trifluoromethyl)thiazole-5-carboxylic acid ethyl ester
72850-79-4
MFCD08166749

Specifications

Ethyl 2-bromo-4-(trifluoromethyl)thiazole-5-carboxylate
Solid
Off-white
304.08
2-Bromo-4-(trifluoromethyl)thiazole-5-carboxylic acid ethyl ester
72850-79-4
97%
5g
MFCD08166749
Videos
SDS