missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-aminopyridine-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number p-7053679
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
13362-30-6
C8H10N2O2
166.18
MFCD03791260
XVBZFXZNJAFCHL-UHFFFAOYSA-N
ethyl 2-aminoisonicotinate, 2-amino-isonicotinic acid ethyl ester, ethyl 2-amino-4-pyridinecarboxylate, 2-aminoisonicotinic acid ethyl ester, 2-amino-4-ethoxycarbonyl pyridine, 4-pyridinecarboxylic acid, 2-amino-, ethyl ester, 2-amino-4-pyridinecarboxylic acid ethyl ester, 2-aminopyridine-4-carboxylic acid ethyl ester, pubchem16599, acmc-209bt6
7015521
ethyl 2-aminopyridine-4-carboxylate
CCOC(=O)C1=CC(=NC=C1)N
This item is not returnable. View return policy
13362-30-6
C8H10N2O2
166.18
MFCD03791260
XVBZFXZNJAFCHL-UHFFFAOYSA-N
ethyl 2-aminoisonicotinate, 2-amino-isonicotinic acid ethyl ester, ethyl 2-amino-4-pyridinecarboxylate, 2-aminoisonicotinic acid ethyl ester, 2-amino-4-ethoxycarbonyl pyridine, 4-pyridinecarboxylic acid, 2-amino-, ethyl ester, 2-amino-4-pyridinecarboxylic acid ethyl ester, 2-aminopyridine-4-carboxylic acid ethyl ester, pubchem16599, acmc-209bt6
7015521
ethyl 2-aminopyridine-4-carboxylate
CCOC(=O)C1=CC(=NC=C1)N

Ethyl 2-aminopyridine-4-carboxylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-aminopyridine-4-carboxylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Color White
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Ethyl 2-aminopyridine-4-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'