missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-aminobenzothiazole-6-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number 15462429
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
50850-93-6
C10H10N2O2S
222.262
MFCD00102724
VYJSGJXWKSDUSG-UHFFFAOYSA-N
601008
ethyl 2-amino-1,3-benzothiazole-6-carboxylate
CCOC(=O)C1=CC2=C(C=C1)N=C(S2)N
This item is not returnable. View return policy
50850-93-6
C10H10N2O2S
222.262
MFCD00102724
VYJSGJXWKSDUSG-UHFFFAOYSA-N
601008
ethyl 2-amino-1,3-benzothiazole-6-carboxylate
CCOC(=O)C1=CC2=C(C=C1)N=C(S2)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 241°C to 245°C
Quantity 5 g
Beilstein 185097
Synonym ethyl 2-aminobenzo d thiazole-6-carboxylate, ethyl 2-amino-benzothiazole-6-carboxylate, ethyl 2-aminobenzothiazole-6-carboxylate, 2-amino-benzothiazole-6-carboxylic acid ethyl ester, 6-benzothiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-6-ethoxycarbonylbenzothiazole, ethyl2-amino-benzothiazole-6-carboxylate, 2-aminobenzothiazole-6-carboxylic acid ethyl ester, 2-amino benzo thiazole-6-carboxylic acid ethyl ester, 3kid
Formula Weight 222.26
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 2-aminobenzothiazole-6-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'