missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-(4-oxo-4H-chromen-3-yl)thiazolidine-4-carboxylate, 97%

Brand:  Alfa Aesar™ H62201.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15437156

  • £398.56 / 5g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-(4-oxo-4H-chromen-3-yl)thiazolidine-4-carboxylate
305.35
MFCD26967913
2-(4-Oxo-4H-chromen-3-yl)thiazolidine-4-carboxylic acid ethyl ester

Specifications

Ethyl 2-(4-oxo-4H-chromen-3-yl)thiazolidine-4-carboxylate
Solid
305.35
2-(4-Oxo-4H-chromen-3-yl)thiazolidine-4-carboxylic acid ethyl ester
5g
97%
MFCD26967913
Videos
SDS