missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-dioxo-4-(2-thienyl)butanoate, 90%, Maybridge™

Brand:  Maybridge SPB04027EA

Code : 31

Additional Details : CAS Number : 36983-36-5 Weight : 0.06000kg

Packaging Amber Glass Bottle
Quantity 10g

 View more versions of this product

Product Code. 10069491

  • £86.00 / 10g

Estimated Shipment: 08-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

36983-36-5
226.246
GCCFYXYKJZUHMI-UHFFFAOYSA-N
2799459
CCOC(=O)C(=O)CC(=O)C1=CC=CS1
C10H10O4S
MFCD00126316
ethyl 2,4-dioxo-4-thiophen-2-yl butanoate, ethyl 2,4-dioxo-4-2-thienyl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid ethyl ester, 2-thiophenebutanoicacid, a,g-dioxo-, ethyl ester, 2,4-dioxo-4-2-thienyl butanoic acid ethyl ester, 2,4-dioxo-4-thiophen-2-ylbutyric acid ethyl ester
ethyl 2,4-dioxo-4-thiophen-2-ylbutanoate

Specifications

Ethyl 2, 4-dioxo-4-(2-thienyl)butanoate
10g
90.0
C10H10O4S
ethyl 2,4-dioxo-4-thiophen-2-yl butanoate, ethyl 2,4-dioxo-4-2-thienyl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid ethyl ester, 2-thiophenebutanoicacid, a,g-dioxo-, ethyl ester, 2,4-dioxo-4-2-thienyl butanoic acid ethyl ester, 2,4-dioxo-4-thiophen-2-ylbutyric acid ethyl ester
CCOC(=O)C(=O)CC(=O)C1=CC=CS1
226.246
226.25
Amber Glass Bottle
36983-36-5
90%
MFCD00126316
GCCFYXYKJZUHMI-UHFFFAOYSA-N
ethyl 2,4-dioxo-4-thiophen-2-ylbutanoate
2799459
90%
Videos
SDS Documents
Certificates