missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-(4-bromophenyl)thiazolidine-4-carboxylate, 97%

Brand:  Alfa Aesar™ H62857.03

Code : NEW

Additional Details : CAS Number : 78979-48-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447926

  • £70.28 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-(4-bromophenyl)thiazolidine-4-carboxylate
316.21
2-(4-Bromophenyl)thiazolidine-4-carboxylic acid ethyl ester
78979-48-3
MFCD27976890

Specifications

Ethyl 2-(4-bromophenyl)thiazolidine-4-carboxylate
Solid
1g
MFCD27976890
78979-48-3
97%
316.21
2-(4-Bromophenyl)thiazolidine-4-carboxylic acid ethyl ester
Videos
SDS