missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Ethyl 2-(3-pentyl)thiazolidine-4-carboxylate, 97%
Brand: Alfa Aesar™ H62988.06
Code : NEW
Additional Details : Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
Ethyl 2-(3-pentyl)thiazolidine-4-carboxylate | |
231.35 | |
MFCD26967921 | |
2-(3-Pentyl)thiazolidine-4-carboxylic acid ethyl ester |
Specifications
Ethyl 2-(3-pentyl)thiazolidine-4-carboxylate | |
5g | |
97% | |
MFCD26967921 |
Colorless | |
Liquid | |
231.35 | |
2-(3-Pentyl)thiazolidine-4-carboxylic acid ethyl ester |