missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-(3-nitrophenyl)thiazolidine-4-carboxylate, 97%

Brand:  Alfa Aesar™ H62125.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15437066

  • £498.20 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-(3-nitrophenyl)thiazolidine-4-carboxylate
282.32
MFCD26967912
2-(3-Nitrophenyl)thiazolidine-4-carboxylic acid ethyl ester

Specifications

Ethyl 2-(3-nitrophenyl)thiazolidine-4-carboxylate
Solid
282.32
2-(3-Nitrophenyl)thiazolidine-4-carboxylic acid ethyl ester
5g
97%
MFCD26967912
Videos
SDS