missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-(3-hydroxyphenyl)thiazolidine-4-carboxylate, 97%

Brand:  Alfa Aesar™ H62592.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15477606

  • £56.92 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-(3-hydroxyphenyl)thiazolidine-4-carboxylate
253.32
MFCD26793812
2-(3-Hydroxyphenyl)thiazolidine-4-carboxylic acid ethyl ester

Specifications

Ethyl 2-(3-hydroxyphenyl)thiazolidine-4-carboxylate
97%
MFCD26793812
1g
Solid
253.32
2-(3-Hydroxyphenyl)thiazolidine-4-carboxylic acid ethyl ester
Videos
SDS