missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-(3-hydroxyphenyl)thiazolidine-4-carboxylate, 97%

Brand:  Alfa Aesar™ H62592.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15487606

  • £285.14 / 5g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-(3-hydroxyphenyl)thiazolidine-4-carboxylate
253.32
MFCD26793812
2-(3-Hydroxyphenyl)thiazolidine-4-carboxylic acid ethyl ester

Specifications

Ethyl 2-(3-hydroxyphenyl)thiazolidine-4-carboxylate
97%
MFCD26793812
5g
Solid
253.32
2-(3-Hydroxyphenyl)thiazolidine-4-carboxylic acid ethyl ester
Videos
SDS