missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-(3,4-difluorophenyl)-4-methylthiazole-5-carboxylate,97%

Brand:  Alfa Aesar™ H58064.04

Code : NEW

Additional Details : Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 15473946

  • £223.55 / 2g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-(3,4-difluorophenyl)-4-methylthiazole-5-carboxylate
283.29
MFCD22581240
2-(3,4-Difluorophenyl)-4-methylthiazole-5-carboxylic acid ethyl ester

Specifications

Ethyl 2-(3,4-difluorophenyl)-4-methylthiazole-5-carboxylate
Pale brown
97%
MFCD22581240
2g
Solid
283.29
2-(3,4-Difluorophenyl)-4-methylthiazole-5-carboxylic acid ethyl ester
Videos
SDS