missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-(3,4-dichlorophenyl)-2-oxoacetate, Technical, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
25 g
Specifications
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 247.08 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | Ethyl 2-(3, 4-dichlorophenyl)-2-oxoacetate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'