missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-[3-(2-indolyl)phenyl]-4-methylthiazole-5-carboxylate, 97%

Brand:  Alfa Aesar™ H58873.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15494606

  • £210.41 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-[3-(2-indolyl)phenyl]-4-methylthiazole-5-carboxylate
363.43
MFCD22581246
2-[3-(2-Indolyl)phenyl]-4-methylthiazole-5-carboxylic acid ethyl ester

Specifications

Ethyl 2-[3-(2-indolyl)phenyl]-4-methylthiazole-5-carboxylate
97%
MFCD22581246
Pale brown
1g
Solid
363.43
2-[3-(2-Indolyl)phenyl]-4-methylthiazole-5-carboxylic acid ethyl ester
260° to 265°C
Videos
SDS