missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-(2-methoxyphenyl)thiazolidine-4-carboxylate, 97%

Brand:  Alfa Aesar™ H62036.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15446956

  • £597.84 / 5g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-(2-methoxyphenyl)thiazolidine-4-carboxylate
267.34
MFCD26967911
2-(2-Methoxyphenyl)thiazolidine-4-carboxylic acid ethyl ester

Specifications

Ethyl 2-(2-methoxyphenyl)thiazolidine-4-carboxylate
5g
97%
MFCD26967911
Yellow
Liquid
267.34
2-(2-Methoxyphenyl)thiazolidine-4-carboxylic acid ethyl ester
Videos
SDS