missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-(2-furyl)thiazolidine-4-carboxylate, 97%

Brand:  Alfa Aesar™ H62161.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458728

  • £597.84 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-(2-furyl)thiazolidine-4-carboxylate
227.28
MFCD27665225
2-(2-Furyl)thiazolidine-4-carboxylic acid ethyl ester

Specifications

Ethyl 2-(2-furyl)thiazolidine-4-carboxylate
97%
MFCD27665225
Pale brown
Liquid
227.28
2-(2-Furyl)thiazolidine-4-carboxylic acid ethyl ester
5g
Videos
SDS