Learn More
Thermo Scientific Maybridge Ethyl 2-(2-formylamino-1,3-thiazol-4-yl)-2-oxoacetate, 1g, 97%
Brand: Thermo Scientific Maybridge SB01141DA
Ethyl 2-(2-formylamino-1,3-thiazol-4-yl)-2-oxoacetate, 1g, 97%
Code : 31
Additional Details : CAS Number : 64987-03-7 Weight : 0.05100kg