missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1-methyl-1H-benzimidazole-5-carboxylate, 97%, Thermo Scientific™
Click to view available options
Amber Glass Bottle; 5g
Specifications
CAS | 53484-19-8 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C11H12N2O2 |
MDL Number | MFCD04501518 |
Quantity | 5 g |
Synonym | ethyl 1-methyl-1h-benzimidazole-5-carboxylate, ethyl 1-methyl-1h-benzo d imidazole-5-carboxylate, ethyl 1-methyl-1,3-benzodiazole-5-carboxylate, 1-methyl-5-benzimidazolecarboxylic acid ethyl ester, 1h-benzimidazole-5-carboxylic acid, 1-methyl-, ethyl ester, 1h-benzimidazole-5-carboxylic acid,1-methyl-,ethyl ester, 1h-benzimidazole-5-carboxylicacid, 1-methyl-, ethyl ester |
InChI Key | MQBXKIFEKNCNJX-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'