missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 1-indoleacetate, 97%

Brand:  Alfa Aesar™ H63518.03

Code : NEW

Additional Details : CAS Number : 61155-69-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15430259

  • £59.78 / 1g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 1-indoleacetate
203.24
1-Indoleacetic acid ethyl ester
61155-69-9
4425801

Specifications

Ethyl 1-indoleacetate
Viscous Liquid
203.24
1-Indoleacetic acid ethyl ester
61155-69-9
97%
4425801
1g
Videos
SDS