missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylate, 97%, Thermo Scientific™

Catalog Number p-3586837
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
926921-61-1
C13H19N3O2
249.31
MFCD09817500
MTWGDARCNWTWJN-UHFFFAOYNA-N
ethyl 1-6-methylpyrazin-2-yl piperidine-3-carboxylate
24229600
ethyl 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylate
CCOC(=O)C1CCCN(C1)C1=NC(C)=CN=C1
This item is not returnable. View return policy
926921-61-1
C13H19N3O2
249.31
MFCD09817500
MTWGDARCNWTWJN-UHFFFAOYNA-N
ethyl 1-6-methylpyrazin-2-yl piperidine-3-carboxylate
24229600
ethyl 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylate
CCOC(=O)C1CCCN(C1)C1=NC(C)=CN=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 249.31
Percent Purity 97%
Chemical Name or Material Ethyl 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'