missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 1,5-dimethyl-1H-pyrazole-3-carboxylate, 97%

Brand:  Alfa Aesar™ H61579.06

Code : NEW

Additional Details : CAS Number : 5744-51-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15428138

  • £80.77 / 5g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 1,5-dimethyl-1H-pyrazole-3-carboxylate
168.19
1,5-Dimethyl-1H-pyrazole-5-carboxylic acid ethyl ester
5744-51-4
MFCD00085071

Specifications

Ethyl 1,5-dimethyl-1H-pyrazole-3-carboxylate
5g
Solid
168.19
1,5-Dimethyl-1H-pyrazole-5-carboxylic acid ethyl ester
Moisture & Light Sensitive
5744-51-4
97%
MFCD00085071
Videos
SDS