missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1,3-dithiolane-2-carboxylate, 99%, Thermo Scientific™

Catalog Number p-128418
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
20461-99-8
C6H10O2S2
178.26
MFCD00005411
OMCSHTHLIQOHDD-UHFFFAOYSA-N
2-carboethoxydithiolane, ethyl1,3-dithiolane-2-carboxylate, 1,3-dithiolane-2-carboxylic acid ethyl ester, acmc-209faj, 2-carboethoxy-1,3-dithiolane, omcshthliqohdd-uhfffaoysa, 1,3-dithiolane-2-carboxylicacid,ethyl ester, 1,3-dithiolane-2-carboxylicacid, ethyl ester, 1,3-dithiolane-2-carboxylic acid, ethyl ester, ethyl 1,3-dithiolane-2-carboxylate
88551
ethyl 1,3-dithiolane-2-carboxylate
CCOC(=O)C1SCCS1
This item is not returnable. View return policy
20461-99-8
C6H10O2S2
178.26
MFCD00005411
OMCSHTHLIQOHDD-UHFFFAOYSA-N
2-carboethoxydithiolane, ethyl1,3-dithiolane-2-carboxylate, 1,3-dithiolane-2-carboxylic acid ethyl ester, acmc-209faj, 2-carboethoxy-1,3-dithiolane, omcshthliqohdd-uhfffaoysa, 1,3-dithiolane-2-carboxylicacid,ethyl ester, 1,3-dithiolane-2-carboxylicacid, ethyl ester, 1,3-dithiolane-2-carboxylic acid, ethyl ester, ethyl 1,3-dithiolane-2-carboxylate
88551
ethyl 1,3-dithiolane-2-carboxylate
CCOC(=O)C1SCCS1
Color Yellow
Density 1.2400g/mL
Boiling Point 85°C (0.1 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5380 to 1.54
Quantity 5 mL
Beilstein 19, II, 290
Specific Gravity 1.24
Formula Weight 178.26
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl 1, 3-dithiolane-2-carboxylate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'