missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 1,3-dimethyl-1H-pyrazole-5-carboxylate, 95%

Brand:  Alfa Aesar™ H61629.03

Code : NEW

Additional Details : CAS Number : 5744-40-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15488178

  • £41.87 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 1,3-dimethyl-1H-pyrazole-5-carboxylate
168.19
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid ethyl ester
5744-40-1
MFCD00233452

Specifications

Ethyl 1,3-dimethyl-1H-pyrazole-5-carboxylate
1g
Pale yellow
95%
MFCD00233452
Moisture & Light Sensitive
5744-40-1
Liquid
168.19
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid ethyl ester
Videos
SDS