missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 1-(2-aminoethyl)piperidine-4-carboxylate dihydrochloride, 97%

Brand:  Alfa Aesar™ H63520.03

Code : NEW

Additional Details : CAS Number : 1177093-16-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15468346

  • £78.86 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 1-(2-aminoethyl)piperidine-4-carboxylate dihydrochloride
273.2
1-(2-aminoethyl)piperidine-4-carboxylic acid ethyl ester dihydrochloride
1177093-16-1
MFCD12022630

Specifications

Ethyl 1-(2-aminoethyl)piperidine-4-carboxylate dihydrochloride
1177093-16-1
97%
MFCD12022630
1g
Solid
273.2
1-(2-aminoethyl)piperidine-4-carboxylic acid ethyl ester dihydrochloride
Videos
SDS