missing translation for 'onlineSavingsMsg'
Learn More

Ethoxycarbonyl isothiocyanate, 95%, Thermo Scientific Chemicals

Catalog Number p-4089830
24.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
16182-04-0
C4H5NO2S
131.15
MFCD00004814
BDTDECDAHYOJRO-UHFFFAOYSA-N
ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate
85320
ethyl N-(sulfanylidenemethylidene)carbamate
CCOC(=O)N=C=S
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
16182-04-0
C4H5NO2S
131.15
MFCD00004814
BDTDECDAHYOJRO-UHFFFAOYSA-N
ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate
85320
ethyl N-(sulfanylidenemethylidene)carbamate
CCOC(=O)N=C=S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.1600g/mL
Boiling Point 56.0°C (18.0 mmHg)
Flash Point 50°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula SCNCOOCH2CH3
Quantity 5 g
Beilstein 03, III, 279
Fieser 04,223; 06,250; 07,146
Specific Gravity 1.16
Formula Weight 131.15
Percent Purity 95%
Chemical Name or Material Ethoxycarbonyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'