missing translation for 'onlineSavingsMsg'
Learn More

Ethoxycarbonyl isocyanate, 90%, tech., Thermo Scientific Chemicals

Catalog Number p-4510539
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19617-43-7
C4H5NO3
115.09
VMVZGGPZNHFGKS-UHFFFAOYSA-N
ethoxycarbonyl isocyanate, ethyl isocyanatoformate, ethyl carbonisocyanatidate, ethyl n-carbonylcarbamate, ethoxycarbonylisocyanate, carbonisocyanatidic acid, ethyl ester, ethyl isocyanatooate, ethylisocyanatoformate, acmc-20al2o, carbonisocyanatidicacid,ethylester
300166
ethyl N-(oxomethylidene)carbamate
CCOC(=O)N=C=O
This item is not returnable. View return policy
19617-43-7
C4H5NO3
115.09
VMVZGGPZNHFGKS-UHFFFAOYSA-N
ethoxycarbonyl isocyanate, ethyl isocyanatoformate, ethyl carbonisocyanatidate, ethyl n-carbonylcarbamate, ethoxycarbonylisocyanate, carbonisocyanatidic acid, ethyl ester, ethyl isocyanatooate, ethylisocyanatoformate, acmc-20al2o, carbonisocyanatidicacid,ethylester
300166
ethyl N-(oxomethylidene)carbamate
CCOC(=O)N=C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.1100g/mL
Boiling Point 25.0°C (10.0 mmHg)
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O2CNCO
Refractive Index 1.4060 to 1.4090
Quantity 1 g
Specific Gravity 1.11
Formula Weight 115.09
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Ethoxycarbonyl isocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'