missing translation for 'onlineSavingsMsg'
Learn More

Ethidium bromide soln., 0.625mg/ml, Thermo Scientific Chemicals

Catalog Number 15444489
Click to view available options
Quantity:
5 mL

 

For visualization of DNA in gel electrophoresis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Red
CAS 1239-45-8
Molecular Formula C21H20BrN3
MDL Number MFCD00011724
UN Number UN2810
Concentration 0.625mg/ml
Merck Index 14,4731
Synonym ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
InChI Key ZMMJGEGLRURXTF-UHFFFAOYSA-N
SMILES [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
IUPAC Name 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
Molecular Weight (g/mol) 394.32
PubChem CID 14710
ChEBI CHEBI:4883
Formula Weight 394.31
Odor Odorless
Quantity 5 mL
Physical Form Dropper Bottle
Chemical Name or Material Ethidium bromide soln.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'