missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethidium Bromide, Fisher BioReagents
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
Description
- Ethidium bromide (EtBr) commonly used as a nucleic acid stain for gel electrophoresis due to its fluorescence when exposed to UV light
- Ethidium bromide intercalates double-stranded DNA and RNA
- Can be used to stain gel after electrophoresis or incorporated into the gel and running buffer
- EtBr also acts as inhibitor of RNA and DNA polymerase
Specifications
Melting Point | 260°C |
Color | Purple |
pH | 4.0 to 7.0 |
Loss on Drying | 5% max. |
Packaging | Amber Glass |
Quantity | 5 g |
Merck Index | 15, 4769 |
Formula Weight | 394.32 |
Physical Form | Powder or Solid |
Chemical Name or Material | Ethidium Bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'