missing translation for 'onlineSavingsMsg'
Learn More

Ethidium bromide, 95%, pure, Thermo Scientific Chemicals

Catalog Number p-4510550
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1239-45-8
C21H20BrN3
394.32
MFCD00011724
ZMMJGEGLRURXTF-UHFFFAOYSA-N
ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
14710
CHEBI:4883
5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
This item is not returnable. View return policy
1239-45-8
C21H20BrN3
394.32
MFCD00011724
ZMMJGEGLRURXTF-UHFFFAOYSA-N
ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
14710
CHEBI:4883
5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 260.0°C to 262.0°C
Color Purple to Red
pH ∼4.5 (1% aq. soln. at 20°C)
Flash Point >100°C
Loss on Drying 5.0% max.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,4769
Solubility Information (1% in water) Clear red
Formula Weight 394.3
Percent Purity 95%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Ethidium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'