missing translation for 'onlineSavingsMsg'
Learn More

Ether USP, Macron Fine Chemicals™

macron

Brand:  Macron Fine Chemicals 0812-16

Additional Details : Weight : 2.50000kg Transport : UN number : 1155 Chem class : 3 Pack group : I

Packaging Al Bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 15597864

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

60-29-7
74.123
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC
C4H10O
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane

Specifications

Ether
C4H10O
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
Al Bottle
60-29-7
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CCOCC
74.123
CHEBI:35702
BP/EP/FCC/JP/NF/USP
2.5kg
Videos
SDS
Documents