missing translation for 'onlineSavingsMsg'
Learn More

Ether USP, Macron Fine Chemicals™

macron

Brand:  Macron Fine Chemicals 0812-10

Additional Details : Weight : 0.50000kg Transport : UN number : 1155 Chem class : 3 Pack group : I

Packaging Al Bottle
Quantity 500g

 View more versions of this product

Product Code. 15587864

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

60-29-7
74.123
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC
C4H10O
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane

Specifications

Ether
C4H10O
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
Al Bottle
60-29-7
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CCOCC
74.123
CHEBI:35702
BP/EP/FCC/JP/NF/USP
500g
Videos
Documents