missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ether USP, Macron Fine Chemicals™
Click to view available options
Quantity:
6 x 500 g
Packaging:
Aluminum Bottle
Specifications
Formula Weight | 74.12 |
Grade | BP/EP/FCC/JP/NF/USP |
Quantity | 6 x 500 g |
Packaging | Aluminum Bottle |
Chemical Name or Material | Ether |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'