missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ether, Preserved with about 2% Alcohol, 0.2% max Water and 7ppm min BHT, U.S.P., J.T. Baker™
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Aluminum Bottle
Specifications
Flash Point | −45.00°C |
Packaging | Aluminum Bottle |
Quantity | 1 L |
Specific Gravity | 1L = 0.71kg |
Formula Weight | 74.12 |
Grade | USP |
Chemical Name or Material | Ether, Preserved with about 2% Alcohol, 0.2% max Water and 7ppm min BHT |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'