missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, purum, 96%, Honeywell™

Brand:  Honeywell Research Chemicals 02884-25L

Code : NEW

Additional Details : CAS Number : 64-17-5 Weight : 20.17500kg Transport : UN number : 1170 Chem class : 1 Pack group : II

Quantity 25L

 View more versions of this product

Product Code. 15640650

  • £2461.61 / 25 L


Estimated Shipment
08-05-2018
Add to basket

Specifications

Chemical Identifiers

44.6mmHg (20°C)
0.789g/mL at 25°C
14°C (57.2°F)
CH3CH2OH
UN1170
46.07g/mol
purum
64-17-5
1.59 (vs air)
19% (60°F)
683°F
MFCD00003568
96%
Fine Spirit
Ethanol
1718733

Specifications

Ethanol
purum
96%
46.07g/mol
UN1170
0.789g/mL at 25°C
683°F
1.59 (vs air)
25L
64-17-5
Fine Spirit
CH3CH2OH
MFCD00003568
1718733
19% (60°F)
44.6mmHg (20°C)
14°C (57.2°F)
Videos
SDS