Ethanol, denatured, special, Alfa Aesar™


Overview
Brand: Alfa Aesar

Manufacturer Part Number: 041603.K7

UNSPSC: 12191601

Code: NEW

Additional Details:
Additional Details: CAS Number: 64-17-5 Weight: 4.00000kg


Quantity 4L

Product Code. 15431717

Quantity Price
1 £ 262.88 / 4 L
Estimated Shipment date
from Supplier 11-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethanol
CAS 64-17-5
CAS 67-56-1
CAS 141-78-6
Physical Form Liquid
Molecular Formula C2H6O
Formula Weight 46.08
MDL Number MFCD00003568

Specification

Color Clear colorless
Physical Form Liquid
Formula Weight 46.08
Quantity 4L
DOT Information DOT Class: 3, Packaging Group: II
Concentration or Composition (by Analyte or Components) Ethyl alcohol: 92%; Methanol: 4%; Ethyl acetate: 4%
Appearance Colorless liquid
MDL Number MFCD00003568
Chemical Name or Material Ethanol
CAS 67-56-1
CAS 64-17-5
Health Hazard 1 H225-H371-H333: Highly flammable liquid and vapour. May cause damage to organs. May be harmful if inhaled.
Odor Alcohol-like
CAS 141-78-6
Molecular Formula C2H6O
EINECS Number 200-578-6
Documentation

Technical documents

Safety and Handling

H225-H371-H333: Highly flammable liquid and vapour. May cause damage to organs. May be harmful if inhaled.