Alfa Aesar™ Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v/v)


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 045844.K2

UNSPSC: 12191601

Code: NEW

Additional Details:
Additional Details: CAS Number: 64-17-5 Weight: 0.78900kg Transport: UN number : 1170 Chem class : 3 Pack group : II


Quantity 1L

Product Code. 15474995

Quantity Price
1 £ 17.490000000000002 / 1 L
Estimated Shipment date
from Supplier 06-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethanol
CAS 64-17-5
Name Note Denatured
Formula Weight 46.08
MDL Number MFCD00003568
UN Number UN1170
Beilstein 1718733
Merck Index 14,3760
Synonym Ethyl alcohol

Specification

Formula Weight 46.08
Health Hazard 3 GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
Name Note Denatured
Quantity 1L
Beilstein 1718733
Merck Index 14,3760
Synonym Ethyl alcohol
Chemical Name or Material Ethanol
MDL Number MFCD00003568
TSCA TSCA
Health Hazard 1 Danger
Assay 91.6%
Density 0.789g/mL
RTECS Number KQ6300000
UN Number UN1170
Color Colorless
Melting Point −130°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
Boiling Point 78°C
Health Hazard 2 GHS H Statement
H225-H361-H371
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
May cause damage to organs.
Refractive Index 1.3610
Physical Form Liquid
Avg. Mol. Wt. or Mol. Wt. Range 1.9% MIBK
Flash Point 8°C (46°F)
CAS 64-17-5
Concentration 3.7% methanol, 1% ethyl acetate, 1% toluene (v/v)
Documentation

Technical documents

Safety and Handling

Danger

GHS H Statement
H225-H361-H371
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
May cause damage to organs.

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.