Alfa Aesar™ Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v/v)


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 045844.M1

1LT Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v

UNSPSC: 12191601

Code: NEW

Additional Details:
Additional Details: CAS Number: 64-17-5 Weight: 1.00000kg


Quantity 4 x 1L

Product Code. 15494995

Quantity Price
1 £ 56.39 / 1 L
Estimated Shipment date
from Supplier 07-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethanol
CAS 64-17-5
Name Note Denatured
Formula Weight 46.08
MDL Number MFCD00003568
UN Number UN1170
Beilstein 1718733
Merck Index 14,3760
Synonym Ethyl alcohol

Specification

Formula Weight 46.08
Health Hazard 3 GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Ethanol
TSCA TSCA
Refractive Index 1.3610
Name Note Denatured
Flash Point 8°C (46°F)
Boiling Point 78°C
Physical Form Liquid
Color Colorless
Melting Point −130°C
Quantity 4 x 1L
Sensitivity Hygroscopic
Avg. Mol. Wt. or Mol. Wt. Range 1.9% MIBK
Assay 91.6%
MDL Number MFCD00003568
CAS 64-17-5
UN Number UN1170
Beilstein 1718733
Merck Index 14,3760
Synonym Ethyl alcohol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H361-H371
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
May cause damage to organs.
Concentration 3.7% methanol, 1% ethyl acetate, 1% toluene (v/v)
Density 0.789g/mL
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
RTECS Number KQ6300000
Safety and Handling

Danger

GHS H Statement
H225-H361-H371
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
May cause damage to organs.

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.