missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, CHROMASOLV™ ACS Reagent, Honeywell Riedel-de Haën™

Catalog Number p-7071700
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
46.069
MFCD00003568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
46.069
MFCD00003568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO
Linear Formula CH3CH2OH
UN Number 1170
Beilstein 1718733
Formula Weight 46.07g/mol
Percent Purity 96% (v/v)
Grade CHROMASOLV™ ACS Reagent
Density 0.789g/mL at 25°C
Quantity 500 mL
Explosion Limit 19% (60°F)
Flash Point 14°C (57.2°F)
Ignition Point 683°F
Physical Form Extra Fine Spirit
Vapor Pressure 44.6mmHg (20°C)
Chemical Name or Material Ethanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'