missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, CHROMASOLV™ ACS Reagent, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
Specifications
Linear Formula | CH3CH2OH |
UN Number | 1170 |
Beilstein | 1718733 |
Formula Weight | 46.07g/mol |
Percent Purity | 96% (v/v) |
Grade | CHROMASOLV™ ACS Reagent |
Density | 0.789g/mL at 25°C |
Quantity | 4 x 2.5 L |
Explosion Limit | 19% (60°F) |
Flash Point | 14°C (57.2°F) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'