Ethanol, anhydrous, denatured, Spectrophotometric Grade, 90%, 5% methanol, 5% isopropanol, Alfa Aesar™


Overview
Brand: Alfa Aesar

Manufacturer Part Number: 022931.M6

UNSPSC: 12191601

Code: NEW

Additional Details:
Additional Details: CAS Number: 64-17-5 Weight: 4.00000kg


Quantity 4 x 4L

Product Code. 15414765

Quantity Price
1 £ 294.04 / 4 L
Estimated Shipment date
from Supplier 18-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethanol
CAS 64-17-5
Purity Grade Spectrophotometric grade
Physical Form Liquid
Assay Percent Range 90%
Molecular Formula C2H6O
Formula Weight 46.08
MDL Number MFCD00003568

Specification

Purity Grade Spectrophotometric grade
Formula Weight 46.08
Quantity 4 x 4L
EINECS Number 200-578-6
Physical Form Liquid
MDL Number MFCD00003568
Density 0.789
Assay Percent Range 90%
Chemical Name or Material Ethanol
CAS 64-17-5
Molecular Formula C2H6O
Documentation

Technical documents