Ethanol, anhydrous, denatured, HPLC Grade, 90%, 5% methanol, 5% isopropanol, Alfa Aesar™


Overview
Brand: Alfa Aesar

Manufacturer Part Number: 022930.K2

UNSPSC: 12191601

Code: NEW

Additional Details:
Additional Details: CAS Number: 64-17-5 Weight: 1.00000kg Transport: UN number : 1987 Chem class : 1 Pack group : III


Quantity 1L

Product Code. 15426065

Quantity Price
1 £ 25.65 / 1 L
Estimated Shipment date
from Supplier 15-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethanol
CAS 64-17-5
Purity Grade HPLC Grade
Physical Form Liquid
Assay Percent Range 90%
Molecular Formula C2H6O
Formula Weight 46.08
MDL Number MFCD00003568

Specification

Purity Grade HPLC Grade
Formula Weight 46.08
Quantity 1L
Color Colorless
UN Number 1987
DOT Information DOT Class: 3; Packing Group: II
Density 0.789g/cm3 at 20°C
Physical Form Liquid
MDL Number MFCD00003568
CAS 64-17-5
EINECS Number 200-578-6
Molecular Formula C2H6O
Odor Alcohol-like
Assay Percent Range 90%
Health Hazard 1 H225-H371-H333: Highly flammable liquid and vapour. May cause damage to organs. May be harmful if inhaled.
Chemical Name or Material Ethanol
Vapor Pressure 57 hPa (43mm Hg) at 20°C
Appearance Colorless liquid
Documentation

Technical documents

Safety and Handling

H225-H371-H333: Highly flammable liquid and vapour. May cause damage to organs. May be harmful if inhaled.