Alfa Aesar™ Ethanol, anhydrous, denatured, 85.8%, 13.3% methanol, 0.9% MIBK


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 036642.M4

UNSPSC: 12191601

Code: NEW

Additional Details:
Additional Details: CAS Number: 64-17-5 Weight: 20.00000kg


Quantity 20L

Product Code. 15456165

Quantity Price
1 £ 234.9 / 20 L
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethanol
CAS 64-17-5
Name Note Anhydrous, denatured
Molecular Formula C2H6O
Linear Formula CH3CH2OH
Formula Weight 46.08
MDL Number MFCD00003568
UN Number UN1993
Beilstein 1718733
Merck Index 14,3760
Synonym Ethyl alcohol

Specification

Formula Weight 46.08
Molecular Formula C2H6O
Assay Percent Range 85.8%
Health Hazard 3 GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Assay 85.8%
Solubility Information Miscible in water and most organic solvents
Sensitivity Hygroscopic
TSCA TSCA
Flash Point 11° (52°F)
Physical Form Liquid
Beilstein 1718733
MDL Number MFCD00003568
Density 0.811g/mL
Refractive Index 1.3610
Synonym Ethyl alcohol
Merck Index 14,3760
Boiling Point 80°C
Color Colorless
Quantity 20L
CAS 64-17-5
Health Hazard 2 GHS H Statement
H225-H370
Highly flammable liquid and vapor.
Causes damage to organs.
Concentration 13.3% methanol, 0.9% MIBK
Odor Like alcohol
Health Hazard 1 Danger
Name Note Anhydrous, denatured
Melting Point −100°C
Linear Formula CH3CH2OH
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Chemical Name or Material Ethanol
RTECS Number KQ6300000
UN Number UN1993

  • As a gasoline additive/substitute; octane booster
  • Very mobile liquid solvent
  • Used in lotions, tonics and perfumes

Safety and Handling

Danger

GHS H Statement
H225-H370
Highly flammable liquid and vapor.
Causes damage to organs.

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.