Ethanol, Alcohol Reagent, anhydrous, denatured, ACS, 94 to 96%, Alfa Aesar™


Overview
Brand: Alfa Aesar

Manufacturer Part Number: 033361.K2

UNSPSC: 12191601

Code: NEW

Additional Details:
Additional Details: CAS Number: 64-17-5 Weight: 0.78900kg Transport: UN number : 1987 Chem class : 1 Pack group : II


Quantity 1L

Product Code. 15416115

Quantity Price
1 £ 31.06 / 1 L
Estimated Shipment date
from Supplier 11-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethanol, Alcohol Reagent, anhydrous, denatured
Name Note For ACS analysis; Begins to lose water at 60°, anhydrous at 100°
CAS 64-17-5
Physical Form Liquid
Assay Percent Range 94-96%
Molecular Formula C2H6O
Formula Weight 46.08
MDL Number MFCD00003568

Specification

Melting Point 12°C
Boiling Point 101°C
Formula Weight 46.08
Quantity 1L
Refractive Index 1.4205 to 1.4220
Flash Point 12°C (53.6°F)
EINECS Number 200-578-6
Physical Form Liquid
CAS 64-17-5
Density 0.789g/mL
Assay Percent Range 94-96%
Name Note For ACS analysis; Begins to lose water at 60°, anhydrous at 100°
Molecular Formula C2H6O
MDL Number MFCD00003568
Chemical Name or Material Ethanol, Alcohol Reagent, anhydrous, denatured
Documentation

Technical documents