Ethanol, Alcohol Reagent, anhydrous, denatured, ACS, 94 to 96%, Alfa Aesar™


Overview
Brand: Alfa Aesar

Manufacturer Part Number: 033361.AP

500ML Ethanol, Alcohol Reagent, anhydrous, denatured, ACS, 94-96% 500ml

UNSPSC: 12191601

Code: NEW

Additional Details:
Additional Details: CAS Number: 64-17-5 Weight: 0.40000kg Transport: UN number : 1987 Chem class : 1 Pack group : II


Quantity 500mL

Product Code. 15406115

Quantity Price
1 £ 24.91 / 500mL
Estimated Shipment date
from Supplier 03-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethanol, Alcohol Reagent, anhydrous, denatured
Name Note For ACS analysis; Begins to lose water at 60°, anhydrous at 100°
CAS 64-17-5
Physical Form Liquid
Assay Percent Range 94-96%
Molecular Formula C2H6O
Formula Weight 46.08
MDL Number MFCD00003568

Specification

Formula Weight 46.08
Quantity 500mL
Chemical Name or Material Ethanol, Alcohol Reagent, anhydrous, denatured
EINECS Number 200-578-6
Name Note For ACS analysis; Begins to lose water at 60°, anhydrous at 100°
Physical Form Liquid
Assay Percent Range 94-96%
Molecular Formula C2H6O
MDL Number MFCD00003568
CAS 64-17-5
Density 0.789g/mL